missing translation for 'onlineSavingsMsg'
Learn More
2-Methylnaphtho[1,2-d]thiazole, 98%, Thermo Scientific Chemicals Product Code.: 11495377

2-Methylnaphtho[1,2-d]thiazole, 98%, Thermo Scientific Chemicals

Product Code. 11495377
5 g
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy

Product Code. 11495377

Brand: Thermo Scientific Alfa Aesar B22725.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2682-45-3
Melting Point 95°C to 97°C
Molecular Formula C12H9NS
MDL Number MFCD00004951
Quantity 5 g
Synonym 2-methylnaphtho 1,2-d thiazole, 2-methyl-beta-naphthothiazole, 2-methylnaphtho 1,2-d 1,3 thiazole, naphtho 1,2-d thiazole, 2-methyl, 2-methyl-b-naphthylthiazole, naphth 1,2-d thiazole, 2-methyl, 2-methyl-.beta.-naphthothiazole, 2-methyinaphtho 1,2-d thiazole, 2-methylbenzo e 1,3 benzothiazole
InChI Key OUXMJRMYZCEVKO-UHFFFAOYSA-N
SMILES CC1=NC2=C(S1)C=CC1=CC=CC=C21
IUPAC Name 2-methylbenzo[e][1,3]benzothiazole
Molecular Weight (g/mol) 199.27
PubChem CID 75892
Formula Weight 199.27
Percent Purity 98%
Chemical Name or Material 2-Methylnaphtho[1,2-d]thiazole
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10201578 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22725.06
Lot Number10226640 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22725.06
Lot Number10171859 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22725.06
Lot Number10210205 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22725.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Methylnaphtho[1,2-d]thiazole, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.