missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylsulfinyl)benzoic acid, 97%, Thermo Scientific™

Product Code. 11456260
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19093-34-6
Molecular Formula C8H8O3S
Molecular Weight (g/mol) 184.21
MDL Number MFCD01936003
InChI Key JVYFTEBQXOBPAB-UHFFFAOYNA-N
Synonym 2-methylsulfinyl benzoic acid, 2-methylsulfinyl benzenecarboxylic acid, 2-methanesulfinylbenzoic acid, o-methylsulfinyl benzoic acid, benzoic acid, o-methylsulfinyl, benzoic acid, 2-methylsulfinyl, acmc-20aomr, 2-methanesulfinyl benzoic acid, o-carboxyphenyl methyl sulfoxide, methylsulfinylbenzenecarboxylicacid
PubChem CID 320058
IUPAC Name 2-methylsulfinylbenzoic acid
SMILES CS(=O)C1=CC=CC=C1C(O)=O

Specifications

Melting Point 150°C to 160°C
Quantity 1 g
Formula Weight 184.21
Percent Purity 97%
Chemical Name or Material 2-(Methylsulfinyl)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'