missing translation for 'onlineSavingsMsg'
Learn More
2-Methyltetrahydrofuran, 99+%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™, Thermo Scientific Chemicals Product Code.: 10104243

2-Methyltetrahydrofuran, 99+%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10104243
100 mL
Click to view available options
Quantity:
100 mL
1 L
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Product Code. 10104243

Brand: Thermo Scientific Acros 396631000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-47-9
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00005367
InChI Key JWUJQDFVADABEY-UHFFFAOYSA-N
Synonym 2-methyltetrahydrofuran, methyltetrahydrofuran, tetrahydrosylvan, tetrahydro-2-methylfuran, furan, tetrahydro-2-methyl, furan, 2-methyl-tetrahydro, 2-methyl tetrahydrofuran, 2-methyl-tetrahydrofuran, 2-methyl-tetrahydro-furan, methyl tetrahydrofuran
PubChem CID 7301
IUPAC Name 2-methyloxolane
SMILES CC1CCCO1

Specifications

Melting Point -136.0°C
CAS Min % 99.0
Color Colorless
CAS Max % 100.0
Density 0.8600g/mL
Boiling Point 78.0°C to 80.0°C
Quantity 100 mL
Infrared Spectrum Authentic
Packaging AcroSeal™ Glass Bottle
Physical Form Liquid
Refractive Index 1.4046 to 1.4066
Assay Percent Range 99% min. (GC)
Specific Gravity 0.86
Viscosity 4 mPa.s (25°C)
Merck Index 15, 6198
Solubility Information Solubility in water: 150g/l (25°C)
Formula Weight 86.13
Percent Purity 99+%
Grade Extra Dry over Molecular Sieve
Chemical Name or Material 2-Methyltetrahydrofuran, Over Molecular Sieves, Stabilized
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2534308 Certificate TypeCertificate of Analysis Date11/04/2025 Catalog Number396630010,396631000
Lot Number2499421 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number396631000,396630010
Lot Number2407368 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number396631000,396630010
Lot Number2416215 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number396631000,396630010
Lot Number1993219 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number396631000,396630010
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Methyltetrahydrofuran
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Methyltetrahydrofuran, 99+%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.