missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)-4,5-dihydro-1H-imidazole hydrobromide, Thermo Scientific™

Product Code. 10622253
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 40241-78-9
Molecular Formula C4H9BrN2S
Molecular Weight (g/mol) 197.094
MDL Number MFCD01570110
InChI Key UPISCLUJZRQPPJ-UHFFFAOYSA-N
Synonym 2-methylthio-4,5-dihydro-1h-imidazole hydrobromide, 4,5-dihydro-2-methylthio-1h-imidazole monohydrobromide, 2-methylsulfanyl-4,5-dihydro-1h-imidazole hydrobromide, 2-methylthio-2-imidazoline, bromide, 1h-imidazole,4,5-dihydro-2-methylthio-, hydrobromide 1:1
PubChem CID 2777335
IUPAC Name 2-methylsulfanyl-4,5-dihydro-1H-imidazole;hydrobromide
SMILES CSC1=NCCN1.Br

Specifications

CAS Min % 80.0
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 197.1
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-(Methylthio)-4, 5-dihydro-1H-imidazole hydrobromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.