Learn More
2-Methylvaleryl chloride, 98%, Thermo Scientific Chemicals
32.90 € - 87.50 €
Chemical Identifiers
CAS | 5238-27-7 |
---|---|
Molecular Formula | C14H12N2O3S |
Molecular Weight (g/mol) | 288.32 |
MDL Number | MFCD00013656 |
InChI Key | NADADZQYJBFZGI-FLIBITNWSA-N |
Synonym | 2-methylvaleryl chloride, 2-methylvaleroyl chloride, 2-methylvalerylchloride, .alpha.-methyl valeroyl chloride, acmc-20anpk, 2-methyvaleryl chloride, pubchem21794, 2-methyl valeryl chloride, 2-methyl-valeryl chloride, 2-methyl pentanoyl chloride |
PubChem CID | 107385 |
SMILES | CC1=CC=C(C=C1)C(\O)=C\C(=O)C(=O)NC1=NC=CS1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11347805
|
Thermo Scientific Alfa Aesar
L02725.06 |
5 g |
32.90 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11357805
|
Thermo Scientific Alfa Aesar
L02725.14 |
25 g |
87.50 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5238-27-7 | |
288.32 | |
NADADZQYJBFZGI-FLIBITNWSA-N | |
107385 |
C14H12N2O3S | |
MFCD00013656 | |
2-methylvaleryl chloride, 2-methylvaleroyl chloride, 2-methylvalerylchloride, .alpha.-methyl valeroyl chloride, acmc-20anpk, 2-methyvaleryl chloride, pubchem21794, 2-methyl valeryl chloride, 2-methyl-valeryl chloride, 2-methyl pentanoyl chloride | |
CC1=CC=C(C=C1)C(\O)=C\C(=O)C(=O)NC1=NC=CS1 |
Specifications
5238-27-7 | |
140°C to 144°C | |
Acrid | |
1.4239 | |
UN2920 | |
Moisture sensitive | |
NADADZQYJBFZGI-FLIBITNWSA-N | |
(3Z)-4-hydroxy-4-(4-methylphenyl)-2-oxo-N-(1,3-thiazol-2-yl)but-3-enamide | |
107385 | |
98% |
0.978 | |
37°C (98°F) | |
C14H12N2O3S | |
MFCD00013656 | |
1743993 | |
2-methylvaleryl chloride, 2-methylvaleroyl chloride, 2-methylvalerylchloride, .alpha.-methyl valeroyl chloride, acmc-20anpk, 2-methyvaleryl chloride, pubchem21794, 2-methyl valeryl chloride, 2-methyl-valeryl chloride, 2-methyl pentanoyl chloride | |
CC1=CC=C(C=C1)C(\O)=C\C(=O)C(=O)NC1=NC=CS1 | |
288.32 | |
134.61 | |
2-Methylvaleryl chloride |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
missing translation for 'einecsNumber' : 226-035-3
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only