Learn More
Thermo Scientific™ TheraPure™ 2'-MOE-T Phosphoramidite, 28-400 finish bottle
2'-MOE-T CEP is a phosphoramidite containing 5' DMT and the nucleoside thymidine (T, or 5-me-U) with 2'-O-methoxy-ethyl (MOE) protection, for use in oligonucleotide synthesis.
19.60 € - 39.30 €
Specifications
For Use With (Equipment) | MerMade Synthesizers |
---|---|
Product Type | 2in-MOE-T CEP |
Content And Storage | Store in freezer (at or below -20°C). |
Nucleoside Type | Thymidine (T) |
Reagent Features | 2in-MOE modified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17937122
|
Thermo Scientific™
27-1021-11 |
1 g |
19.60 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17907132
|
Thermo Scientific™
27-1021-82 |
2 g |
39.30 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2'-MOE-T CEP is a phosphoramidite containing 5' DMT and the nucleoside thymidine (T, or 5-me-U) with 2'-O-methoxy-ethyl (MOE) protection, for use in oligonucleotide synthesis. Located in the ideal oligo position, 2'-O-methoxy-ethyl (MOE) modifications display nuclease resistance, fewer off-target effects, and slightly increased hybridization affinities. These phosphoramidites are commonly used for oligos designed for antisense, siRNA, aptamer, or gRNA-based research. The TheraPure trademark denotes our highest level of phosphoramidite purity, developed with the greatest stringency of high definition analytics and tightly controlled impurity profiles and, therefore, ideal for any therapeutic application. This product is supplied in a 30-mL amber bottle with a 28-400 finish neck style, which is compatible with most MerMade synthesizers.
Features:
• Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-2'-O-methoxyethyl-thymidine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula: C43H55N5O10P
• Base: T
• Formula weight: 819.0 g/mol
• CAS number: 163878-63-5
• HPLC purity: ≥99%; M-11 Impurity ≤0.01%
• 31P NMR purity: ≥99%
Specifications
MerMade Synthesizers | |
Store in freezer (at or below -20°C). | |
2in-MOE modified | |
99% | |
TheraPure™ | |
TheraPure™ |
2in-MOE-T CEP | |
Thymidine (T) | |
Room Temperature | |
Phosphoramidite | |
RNA | |
Powder |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.