missing translation for 'onlineSavingsMsg'
Learn More

(2-Morpholino-3-pyridinyl)methanol, 97%, Thermo Scientific™

Product Code. 10581492
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 423768-55-2
Molecular Formula C10H14N2O2
Molecular Weight (g/mol) 194.23
MDL Number MFCD03086196
InChI Key WESHZTWFDUKWGL-UHFFFAOYSA-N
PubChem CID 2776571
IUPAC Name (2-morpholin-4-ylpyridin-3-yl)methanol
SMILES OCC1=C(N=CC=C1)N1CCOCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 2-morpholino-3-pyridinyl methanol, 2-morpholin-4-yl pyridin-3-yl methanol, 3-pyridinemethanol,2-4-morpholinyl, 2-morpholinopyridin-3-yl methanol, 3-pyridinemethanol, 2-4-morpholinyl, 2-morpholin-4-ylpyridin-3-yl methanol, 2-morpholin-4-yl-3-pyridyl methan-1-ol
Formula Weight 194.23
Percent Purity 97%
Chemical Name or Material (2-Morpholino-3-pyridinyl)methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'