missing translation for 'onlineSavingsMsg'
Learn More

2-Morpholino-5-nitrobenzaldehyde, 97%, Thermo Scientific™

Product Code. 10243232
Click to view available options
Amber Glass Bottle; 1g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 30742-62-2
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C11H12N2O4
MDL Number MFCD00439921
Quantity 1 g
Synonym 2-morpholino-5-nitrobenzaldehyde, 2-morpholin-4-yl-5-nitrobenzaldehyde, 2-morpholino-5-nitrobenzenecarbaldehyde, 2-4-morpholinyl-5-nitrobenzaldehyde, 4-2-formyl-4-nitrophenyl morpholine, 2-morpholin-4-yl-5-nitro-benzaldehyde, 3-formyl-4-morpholin-4-yl nitrobenzene, benzaldehyde, 2-4-morpholinyl-5-nitro
InChI Key PCGDWIWUQDHQLK-UHFFFAOYSA-N
SMILES C1COCCN1C2=C(C=C(C=C2)[N+](=O)[O-])C=O
IUPAC Name 2-morpholin-4-yl-5-nitrobenzaldehyde
Molecular Weight (g/mol) 236.227
PubChem CID 2794872
Formula Weight 236.23
Percent Purity 97%
Chemical Name or Material 2-Morpholino-5-nitrobenzaldehyde
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.