missing translation for 'onlineSavingsMsg'
Learn More

2-n-Hexylpyridine, 98%

Product Code. 11445113
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11445113

Brand: Thermo Scientific Alfa Aesar A12772.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1129-69-7 | C11H17N | 163.264 g/mol

2-n-Hexylpyridine is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-n-Hexylpyridine is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Slightly soluble in water.

Notes
This product is relativelyunstable under normal temperature. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1129-69-7
Molecular Formula C11H17N
Molecular Weight (g/mol) 163.264
MDL Number MFCD00023552
InChI Key NZLJDTKLZIMONR-UHFFFAOYSA-N
Synonym pyridine, 2-hexyl, 2-n-hexylpyridine, 2-n-hexyl pyridine, acmc-20antt, pyridine, 2-n-hexyl, 5-20-06-00161 beilstein handbook reference
PubChem CID 70797
IUPAC Name 2-hexylpyridine
SMILES CCCCCCC1=CC=CC=N1

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 163.27
Percent Purity 98%
Chemical Name or Material 2-n-Hexylpyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.