missing translation for 'onlineSavingsMsg'
Learn More

2-n-Propyl-1-heptanol, 98%, Thermo Scientific Chemicals

Product Code. 11439520
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-n-Propyl-1-heptanol is used as a starting material for the production of plasticizers for PVC, e.g. Palatinol™ 10-P.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-n-Propyl-1-heptanol is used as a starting material for the production of plasticizers for PVC, e.g. Palatinol™ 10-P.

Solubility
Insoluble in water. Miscible with most common organic solvents.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10042-59-8
Molecular Formula C10H22O
Molecular Weight (g/mol) 158.285
MDL Number MFCD00046768
InChI Key YLQLIQIAXYRMDL-UHFFFAOYSA-N
Synonym 2-propyl-1-heptanol, 1-heptanol, 2-propyl, 2-propylheptanol, 2-n-propyl-1-heptanol, 2-propylheptyl alcohol, 2-propyl heptanol, 2-propyl-heptan-1-ol, acmc-20ann4, 2-propyl-1-heptanol #, dsstox_cid_9302
PubChem CID 24847
IUPAC Name 2-propylheptan-1-ol
SMILES CCCCCC(CCC)CO

Specifications

Quantity 1 g
Solubility Information Insoluble in water. Miscible with most common organic solvents.
Formula Weight 158.28
Percent Purity 98%
Chemical Name or Material 2-n-Propyl-1-heptanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'