missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-n-Propyl-1-heptanol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
2-n-Propyl-1-heptanol is used as a starting material for the production of plasticizers for PVC, e.g. Palatinol™ 10-P.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-n-Propyl-1-heptanol is used as a starting material for the production of plasticizers for PVC, e.g. Palatinol™ 10-P.
Solubility
Insoluble in water. Miscible with most common organic solvents.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
2-n-Propyl-1-heptanol is used as a starting material for the production of plasticizers for PVC, e.g. Palatinol™ 10-P.
Solubility
Insoluble in water. Miscible with most common organic solvents.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
Quantity | 5 g |
Solubility Information | Insoluble in water. Miscible with most common organic solvents. |
Formula Weight | 158.28 |
Percent Purity | 98% |
Chemical Name or Material | 2-n-Propyl-1-heptanol |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction