Learn More
2-Naphthoyl chloride, 98%, Thermo Scientific Chemicals
54.37 € - 217.08 €
Chemical Identifiers
CAS | 2243-83-6 |
---|---|
Molecular Formula | C11H7ClO |
Molecular Weight (g/mol) | 190.63 |
MDL Number | MFCD00004093 |
InChI Key | XNLBCXGRQWUJLU-UHFFFAOYSA-N |
Synonym | 2-naphthoyl chloride, 2-naphthalenecarbonyl chloride, 2-naphthoic chloride, 2-naphthoylchloride, beta-naphthoyl chloride, 2-napthoyl chloride, 2-chlorocarbonyl naphthalene, .beta.-naphthoyl chloride, .beta.-naphthalenecarbonyl chloride, beta-naphthalenecarbonyl chloride |
PubChem CID | 75246 |
IUPAC Name | naphthalene-2-carbonyl chloride |
SMILES | C1=CC=C2C=C(C=CC2=C1)C(=O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10189439
|
Thermo Scientific Acros
376590100 |
10 g | Glass bottle |
54.37 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10873961
|
Thermo Scientific Acros
376590500 |
50 g | Glass bottle |
217.08 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische Identifikatoren
2243-83-6 | |
190.63 | |
XNLBCXGRQWUJLU-UHFFFAOYSA-N | |
75246 | |
C1=CC=C2C=C(C=CC2=C1)C(=O)Cl |
C11H7ClO | |
MFCD00004093 | |
2-naphthoyl chloride, 2-naphthalenecarbonyl chloride, 2-naphthoic chloride, 2-naphthoylchloride, beta-naphthoyl chloride, 2-napthoyl chloride, 2-chlorocarbonyl naphthalene, .beta.-naphthoyl chloride, .beta.-naphthalenecarbonyl chloride, beta-naphthalenecarbonyl chloride | |
naphthalene-2-carbonyl chloride |
Spezifikation
156.0°C to 158.0°C (7.0mmHg) | |
2243-83-6 | |
Crystalline Chunks | |
C11H7ClO | |
MFCD00004093 | |
2-naphthoyl chloride, 2-naphthalenecarbonyl chloride, 2-naphthoic chloride, 2-naphthoylchloride, beta-naphthoyl chloride, 2-napthoyl chloride, 2-chlorocarbonyl naphthalene, .beta.-naphthoyl chloride, .beta.-naphthalenecarbonyl chloride, beta-naphthalenecarbonyl chloride | |
XNLBCXGRQWUJLU-UHFFFAOYSA-N | |
naphthalene-2-carbonyl chloride | |
75246 | |
98% | |
2-Naphthoyl chloride |
49.0°C to 54.0°C | |
Brown to Gray | |
97.5% min. (GC) | |
C10H7COCl | |
>110°C | |
Solubility in water: decomposes. Other solubilities: soluble in aceton | |
C1=CC=C2C=C(C=CC2=C1)C(=O)Cl | |
190.63 | |
190.63 | |
Authentic |
Sicherheit und Handhabung
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 218-822-5
- 2-Naphthoyl chloride
Signalwort
- Danger
Gefahrenkategorie
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Gefahrenhinweis
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- EUH029-Contact with water liberates toxic gas.
Sicherheitshinweis
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.