missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-1-propanol, 97%, Thermo Scientific Chemicals

Product Code. 11310508
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Nitro-1-propanol is used in improved synthesis of 2,2,2-trinitroethyl ethers, inhibits methane production during in vitro ruminal fermentation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2902-96-7
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.09
MDL Number MFCD00007398
InChI Key PCNWBUOSTLGPMI-UHFFFAOYNA-N
Synonym 2-nitro-1-propanol, 2-nitropropanol, 1-propanol, 2-nitro, 2-nitropropanol-1, ccris 5045, 2-nitro-propan-1-ol, acmc-20anp9, 1-hydroxy-2-nitropropane, 1-propanol, 2-nitro-8ci
PubChem CID 92187
IUPAC Name 2-nitropropan-1-ol
SMILES CC(CO)[N+]([O-])=O

Specifications

Density 1.185 g/mL
Boiling Point 73°C (1 mmHg)
Flash Point 100°C (212°F)
Refractive Index 1.439
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 105.09
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Nitro-1-propanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'