missing translation for 'onlineSavingsMsg'
Learn More
2-Nitro-4-(trifluoromethyl)aniline, 98%, Thermo Scientific Chemicals Product Code.: 11407047

2-Nitro-4-(trifluoromethyl)aniline, 98%, Thermo Scientific Chemicals

Product Code. 11407047
50 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11407047

Brand: Thermo Scientific Alfa Aesar A14413.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 400-98-6
Molecular Formula C7H5F3N2O2
Molecular Weight (g/mol) 206.124
MDL Number MFCD00007155
InChI Key ATXBGHLILIABGX-UHFFFAOYSA-N
Synonym 2-nitro-4-trifluoromethyl aniline, 4-amino-3-nitrobenzotrifluoride, benzenamine, 2-nitro-4-trifluoromethyl, alpha,alpha,alpha-trifluoro-2-nitro-p-toluidine, 4-trifluoromethyl 2-nitroaniline, 4-amino-3-nitro benzotrifluoride, 4-amino-3-nitrobenzo trifluoride, aniline, 2-nitro-4-trifluoromethyl, 2-nitro-4-trifluoromethyl phenylamine, 3-nitro-4-aminobenzotrifluorideShow MoreShow Less
PubChem CID 67865
IUPAC Name 2-nitro-4-(trifluoromethyl)aniline
SMILES C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])N

Specifications

Melting Point 105°C to 109°C
Quantity 50 g
UN Number UN2811
Beilstein 650808
Formula Weight 206.13
Percent Purity 98%
Chemical Name or Material 2-Nitro-4-(trifluoromethyl)aniline

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10150727 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA14413.09,A14413.18,A14413.30
Lot Number10151197 Certificate TypeCertificate of Analysis Date12/01/2024 Catalog NumberA14413.09,A14413.18,A14413.30
Lot Number10241354 Certificate TypeCertificate of Analysis Date20/06/2022 Catalog NumberA14413.09,A14413.18,A14413.30
Lot NumberI3826A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14413.09,A14413.18,A14413.30
Lot Number10099748 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14413.09,A14413.18,A14413.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Nitro-4-(trifluoromethyl)aniline, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.