missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Nitro-p-phenylenediamine, 95%, Thermo Scientific Chemicals
32.20 € - 246.00 €
Chemical Identifiers
CAS | 5307-14-2 |
---|---|
Molecular Formula | C6H7N3O2 |
Molecular Weight (g/mol) | 153.141 |
MDL Number | MFCD00007903 |
InChI Key | HVHNMNGARPCGGD-UHFFFAOYSA-N |
Synonym | 2-nitro-1,4-phenylenediamine, 1,4-diamino-2-nitrobenzene, 2-nitro-p-phenylenediamine, 2,5-diaminonitrobenzene, 1,4-benzenediamine, 2-nitro, durafur brown, 2-nitro-1,4-benzenediamine, o-nitro-p-phenylenediamine, 4-amino-2-nitroaniline, fouramine 2r |
PubChem CID | 4338370 |
ChEBI | CHEBI:76394 |
IUPAC Name | 2-nitrobenzene-1,4-diamine |
SMILES | C1=CC(=C(C=C1N)[N+](=O)[O-])N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11435477
|
Thermo Scientific Alfa Aesar
B23477.14 |
25 g |
32.20 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11445477
|
Thermo Scientific Alfa Aesar
B23477.22 |
100 g |
67.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11455477
|
Thermo Scientific Alfa Aesar
B23477.36 |
500 g |
246.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5307-14-2 | |
153.141 | |
HVHNMNGARPCGGD-UHFFFAOYSA-N | |
4338370 | |
2-nitrobenzene-1,4-diamine |
C6H7N3O2 | |
MFCD00007903 | |
2-nitro-1,4-phenylenediamine, 1,4-diamino-2-nitrobenzene, 2-nitro-p-phenylenediamine, 2,5-diaminonitrobenzene, 1,4-benzenediamine, 2-nitro, durafur brown, 2-nitro-1,4-benzenediamine, o-nitro-p-phenylenediamine, 4-amino-2-nitroaniline, fouramine 2r | |
CHEBI:76394 | |
C1=CC(=C(C=C1N)[N+](=O)[O-])N |
Specifications
5307-14-2 | |
C6H7N3O2 | |
2-nitro-1,4-phenylenediamine, 1,4-diamino-2-nitrobenzene, 2-nitro-p-phenylenediamine, 2,5-diaminonitrobenzene, 1,4-benzenediamine, 2-nitro, durafur brown, 2-nitro-1,4-benzenediamine, o-nitro-p-phenylenediamine, 4-amino-2-nitroaniline, fouramine 2r | |
C1=CC(=C(C=C1N)[N+](=O)[O-])N | |
153.141 | |
CHEBI:76394 | |
95% |
134°C to 137°C | |
MFCD00007903 | |
HVHNMNGARPCGGD-UHFFFAOYSA-N | |
2-nitrobenzene-1,4-diamine | |
4338370 | |
153.14 | |
2-Nitro-p-phenylenediamine |
Safety and Handling
GHS H Statement
H341-H351
Suspected of causing genetic defects.
Suspected of causing cancer.
P201-P202-P281-P308+P313-P501c
H341
missing translation for 'einecsNumber' : 226-164-5
missing translation for 'rtecsNumber' : ST3000000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only