missing translation for 'onlineSavingsMsg'
Learn More
2-Nitroaniline, 98%, Thermo Scientific Chemicals Product Code.: 11428903

2-Nitroaniline, 98%, Thermo Scientific Chemicals

Product Code. 11428903
5000 g
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy

Product Code. 11428903

Brand: Thermo Scientific Alfa Aesar A11055.0I

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Nitroaniline is the main precursor to phenylenediamines, which are converted to benzimidazoles, a family of heterocycles that are key components in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nitroaniline is the main precursor to phenylenediamines, which are converted to benzimidazoles, a family of heterocycles that are key components in pharmaceuticals.

Solubility
Soluble in water (0.6 g/L at 20°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-74-4
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.126
MDL Number MFCD00007687
InChI Key DPJCXCZTLWNFOH-UHFFFAOYSA-N
Synonym o-nitroaniline, benzenamine, 2-nitro, o-nitraniline, 1-amino-2-nitrobenzene, 2-nitrobenzenamine, aniline, o-nitro, devol orange b, orange grs salt, o-aminonitrobenzene, 2-aminonitrobenzene
PubChem CID 6946
IUPAC Name 2-nitroaniline
SMILES C1=CC=C(C(=C1)N)[N+](=O)[O-]

Specifications

Melting Point 70°C to 74°C
Density 1.255
Boiling Point 284°C to 286°C
Flash Point 168°C (334°F)
Quantity 5000 g
UN Number UN1661
Beilstein 509275
Merck Index 14,6583
Solubility Information Soluble in water (0.6g/L at 20°C).
Formula Weight 138.13
Percent Purity 98%
Chemical Name or Material 2-Nitroaniline
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Nitroaniline, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.