missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzaldehyde, 97%, Thermo Scientific™

Product Code. 10799395
Click to view available options
Quantity:
10 g
50 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 552-89-6
Molecular Formula C7H5NO3
Molecular Weight (g/mol) 151.121
MDL Number MFCD00007132
InChI Key CMWKITSNTDAEDT-UHFFFAOYSA-N
Synonym o-nitrobenzaldehyde, benzaldehyde, 2-nitro, nitrobenzaldehyde, benzaldehyde, o-nitro, 2-nitro-benzaldehyde, 2-formylnitrobenzene, ccris 2322, unii-48b18q9b8e, benzaldehyde, nitro, ortho-nitrobenzaldehyde
PubChem CID 11101
ChEBI CHEBI:66927
IUPAC Name 2-nitrobenzaldehyde
SMILES C1=CC=C(C(=C1)C=O)[N+](=O)[O-]

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula O2NC6H4CHO
Quantity 10 g
Formula Weight 151.12
Percent Purity 97%
Chemical Name or Material 2-Nitrobenzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'