Learn More
2-Nitroimidazole, 98%, Thermo Scientific Chemicals
89.80 € - 315.00 €
Chemical Identifiers
CAS | 527-73-1 |
---|---|
Molecular Formula | C3H3N3O2 |
Molecular Weight (g/mol) | 113.08 |
MDL Number | MFCD00005185 |
InChI Key | YZEUHQHUFTYLPH-UHFFFAOYSA-N |
Synonym | 2-nitroimidazole, azomycin, amicin, nitroimidazole, 1h-imidazole, 2-nitro, imidazole, 2-nitro, 2-nitroimidazol, 2-nitro-imidazole, unii-k8e96xl55d, azomycin 2-nitroimidazole |
PubChem CID | 10701 |
ChEBI | CHEBI:67135 |
IUPAC Name | 2-nitro-1H-imidazole |
SMILES | [O-][N+](=O)C1=NC=CN1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
527-73-1 | |
113.08 | |
YZEUHQHUFTYLPH-UHFFFAOYSA-N | |
10701 | |
2-nitro-1H-imidazole |
C3H3N3O2 | |
MFCD00005185 | |
2-nitroimidazole, azomycin, amicin, nitroimidazole, 1h-imidazole, 2-nitro, imidazole, 2-nitro, 2-nitroimidazol, 2-nitro-imidazole, unii-k8e96xl55d, azomycin 2-nitroimidazole | |
CHEBI:67135 | |
[O-][N+](=O)C1=NC=CN1 |
Specifications
527-73-1 | |
100.0 | |
Green-Khaki to Yellow | |
98% | |
MFCD00005185 | |
2-nitroimidazole, azomycin, amicin, nitroimidazole, 1h-imidazole, 2-nitro, imidazole, 2-nitro, 2-nitroimidazol, 2-nitro-imidazole, unii-k8e96xl55d, azomycin 2-nitroimidazole | |
YZEUHQHUFTYLPH-UHFFFAOYSA-N | |
2-nitro-1H-imidazole | |
10701 | |
113.08 | |
Powder |
97.5 | |
287°C | |
Authentic | |
C3H3N3O2 | |
15, 914 | |
Solubility in water: insoluble in water. Other solubilities: soluble in alkaline h2o- insoluble in acidic h2o,soluble in methanol,ethanol,acetone,ethyl,acetate and butyl acetate,practically insoluble in ether and chloroform | |
[O-][N+](=O)C1=NC=CN1 | |
113.08 | |
CHEBI:67135 | |
98% | |
2-Nitroimidazole, 98% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye prot
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 208-425-5
missing translation for 'rtecsNumber' : NI7875000
missing translation for 'tsca' : TSCA
- 2-Nitroimidazole
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H301-Toxic if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only