missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrophenol, 99%, Thermo Scientific Chemicals

Product Code. 10316680
100 g, Glass bottle
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-75-5
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00011688
InChI Key IQUPABOKLQSFBK-UHFFFAOYSA-N
Synonym o-nitrophenol, 2-hydroxynitrobenzene, phenol, 2-nitro, o-hydroxynitrobenzene, phenol, o-nitro, nitrophenol, o-nitrofenol, ortho-nitrophenol, phenol, nitro, o-nitrofenol czech
PubChem CID 6947
ChEBI CHEBI:16260
IUPAC Name 2-nitrophenol
SMILES OC1=CC=CC=C1[N+]([O-])=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 43.0°C to 47.0°C
Color Yellow
Boiling Point 214.0°C to 216.0°C
Flash Point 108°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4OH
Quantity 100 g
UN Number 1663
Beilstein 06, 213
Fieser 05,477
Merck Index 15, 6707
Solubility Information Solubility in water: 2g/L (25°C). Other solubilities: soluble in alcohol,ether,acetone,chloroform,,benzene,carbon disulfide,alkali hydroxides,and toluene
Viscosity 1.35 mPa.s (80°C)
Formula Weight 139.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Nitrophenol, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Nitrophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P273-Avoid release to the environment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Nitrophenol, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.