missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrophenylhydrazine, 97%, moistened with ca 30% water, Thermo Scientific Chemicals

Product Code. 10795401
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3034-19-3, 7732-18-5
CAS Min % 96.0
CAS Max % 100.0
Melting Point 89.0°C to 94.0°C
Color Orange to Red
Loss on Drying 30% to 35%
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C6H7N3O2
Linear Formula O2NC6H4NHNH2
MDL Number MFCD00007577
Quantity 10 g
Synonym 2-nitrophenyl hydrazine, hydrazine, nitrophenyl, hydrazine, o-nitrophenyl, o-nitrophenylhydrazine, hydrazine, 2-nitrophenyl, 2-nitrophenylhrdrazine, 2-nitrophenyl diazane, pubchem12859, acmc-1agti, o-nitrophenyl hydrazine
Solubility Information Solubility in water: miscible. Other solubilities: soluble in benzene
InChI Key FRBUNLLUASHNDJ-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)NN)[N+](=O)[O-]
IUPAC Name (2-nitrophenyl)hydrazine
Molecular Weight (g/mol) 153.14
PubChem CID 3314739
Formula Weight 153.14
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Nitrophenylhydrazine, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Nitrophenylhydrazine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'