missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrotoluene, 99+%, Thermo Scientific Chemicals

Product Code. 10225180
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-72-2
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
InChI Key PLAZTCDQAHEYBI-UHFFFAOYSA-N
Synonym 2-nitrotoluene, o-nitrotoluene, o-nitrotoluol, o-methylnitrobenzene, benzene, 1-methyl-2-nitro, toluene, o-nitro, 2-methyl-1-nitrobenzene, 2-nitrotoluol, ortho-nitrotoluol, 2-methylnitrobenzene
PubChem CID 6944
ChEBI CHEBI:33098
IUPAC Name 1-methyl-2-nitrobenzene
SMILES CC1=CC=CC=C1[N+](=O)[O-]

Specifications

Melting Point -9.0°C
Density 1.1600g/mL
Boiling Point 222.0°C
Flash Point 95°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CH3C6H4NO2
Refractive Index 1.5445 to 1.5465
Quantity 1 L
Beilstein 05,318
Merck Index 15,6737
Specific Gravity 1.16
Solubility Information Solubility in water: 0.44g/L (20°C). Other solubilities: soluble in alcohol, ether and benzene
Formula Weight 137.14
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2-Nitrotoluene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Nitrotoluene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 2
Hazard Statement
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H361f-Suspected of damaging fertility.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'