Learn More
2’-O,4’-C-Methyleneuridine, MedChemExpress
MedChemExpress 2’-O,4’-C-Methyleneuridine (Compound 15a) is a bicyclic nucleoside.
Brand:  Medchem Express HY-111639
Encompass_Preferred
 View more versions of this product
Product Code. 16414508
  • 199.80 € / 10mg
Explore more special offers
Description
Description
2’-O,4’-C-Methyleneuridine (Compound 15a) is a bicyclic nucleoside.
Specifications
Specifications
2’-O,4’-C-Methyleneuridine
White
C10H12N2O6
DMSO : 125 mg/mL (487.88 mM; Need ultrasonic)
256.21
99.49%
Solid
200435-92-3
Research
10 mg
OC[C@@]12O[C@@H](N3C(NC(C=C3)=O)=O)[C@H](OC2)[C@@H]1O
256.21
Research
Neuroscience-Neuromodulation