Learn More
Thermo Scientific™ 2'-OMe-Bz-A Phosphoramidite, standard grade, 28-400 finish bottle
2'-OMe-Bz-A CEP is a phosphoramidite containing 5' DMT and the nucleoside adenosine (A) with 2'-O-methyl protection, for use in oligonucleotide synthesis.
Brand: Thermo Scientific™ 27-1842-11
Description
2'-OMe-Bz-A CEP is a phosphoramidite containing 5' DMT and the nucleoside adenosine (A) with 2'-O-methyl protection, for use in oligonucleotide synthesis. Located in the ideal oligo position, 2'-O-methyl (OMe) oligo modifications display nuclease resistance, fewer off-target effects, and slightly increased hybridization affinities. These phosphoramidites are commonly used for oligos designed for antisense, siRNA, aptamer, or gRNA-based research. The 'standard grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in a 30-mL amber bottle with a 28-400 finish neck style, which is compatible with most MerMade synthesizers.
Features:
• Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-N6-benzoyl-2'-O-methyl-adenosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula: C48H54N7O8P
• Base: A
• Base Protection: Bz
• Formula weight: 887.97 g/mol
• CAS number: 110782-31-5
• HPLC purity: ≥98.0%; Sum of non-primary peaks 140-152 ppm: ≤0.5%
• 31P NMR purity: ≥98.0%

Specifications
MerMade Synthesizers | |
Store in freezer (at or below -20°C). | |
2in-OMe modified | |
98% | |
Phosphoramidite | |
RNA |
2in-OMe-Bz-A CEP | |
Adenosine (A) | |
Room Temperature | |
1 g | |
Standard | |
Powder |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
ZA4552 | Certificate of Analysis | 28/01/2025 | 27-1842-11 |
ZC1752 | Certificate of Analysis | 26/01/2025 | 27-1842-11 |
ZF0352 | Certificate of Analysis | 26/01/2025 | 27-1842-11 |
ZI1152 | Certificate of Analysis | 25/01/2025 | 27-1842-11 |
ZK0642 | Certificate of Analysis | 25/01/2025 | 27-1842-11 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.