missing translation for 'onlineSavingsMsg'
Learn More
2-Oxa-5-azaspiro[3.4]octane oxalate, 96%, Thermo Scientific Chemicals Product Code.: 15484248

2-Oxa-5-azaspiro[3.4]octane oxalate, 96%, Thermo Scientific Chemicals

Product Code. 15484248
500 mg
Click to view available options
Quantity:
250 mg
500 mg
This item is not returnable. View return policy

Product Code. 15484248

Brand: Thermo Scientific Alfa Aesar H57854.ME

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Spiro forms of lactones and oxazines find applications as leuco dyes, by frequently displaying chromism - reversibly interchanging between their colorless and colored forms. Spiro compounds are used as photochromic materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Spiro forms of lactones and oxazines find applications as leuco dyes, by frequently displaying chromism - reversibly interchanging between their colorless and colored forms. Spiro compounds are used as photochromic materials.

Solubility
Decomposes in water.

Notes
Keep container tightly closed. Keep container in a cool refrigerated, well-ventilated area. Moisture sensitive
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1389264-18-9
Molecular Formula C8H13NO5
Molecular Weight (g/mol) 203.194
MDL Number MFCD18325162
InChI Key JFOZNINEJYPQQK-UHFFFAOYSA-N
Synonym 2-oxa-5-azaspiro 3.4 octane oxalate, 2-oxa-5-aza-spiro 3.4 octane oxalate, 2-oxa-5-azaspiro 3.4 octane oxalate salt, 2-oxa-5-azaspiro 3.4 octane, hemioxalate salt, 2-oxa-5-azaspiro 3.4 octane; oxalic acid
PubChem CID 72207353
IUPAC Name 2-oxa-5-azaspiro[3.4]octane;oxalic acid
SMILES C1CC2(COC2)NC1.C(=O)(C(=O)O)O

Specifications

Quantity 500 mg
Solubility Information Decomposes in water.
Formula Weight 203.19
Percent Purity 96%
Chemical Name or Material 2-Oxa-5-azaspiro[3.4]octane oxalate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberD25Z013 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH57854.MD,H57854.ME
Lot NumberX02A007 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH57854.MD,H57854.ME

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Oxa-5-azaspiro[3.4]octane oxalate, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.