missing translation for 'onlineSavingsMsg'
Learn More
2-Oxa-6-azaspiro[3.3]heptane oxalate, 97%, Thermo Scientific Chemicals Product Code.: 15470339

2-Oxa-6-azaspiro[3.3]heptane oxalate, 97%, Thermo Scientific Chemicals

Product Code. 15470339
250 mg
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15470339

Brand: Thermo Scientific Alfa Aesar H63578.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Oxa-6-azaspiro[3.3]heptane oxalate used as a intermediate in pharmaceutical and chemical synthesis. Also used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Oxa-6-azaspiro[3.3]heptane oxalate used as a intermediate in pharmaceutical and chemical synthesis. Also used in medicine.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1159599-99-1
Molecular Formula C7H11NO5
Molecular Weight (g/mol) 189.17
MDL Number MFCD11976178
InChI Key KOUVDKDABFOPIG-UHFFFAOYSA-N
Synonym 2-oxa-6-azaspiro 3.3 heptane oxalate, 2-oxa-6-aza-spiro 3.3 heptane oxalic acid salt, 2-oxa-6-azaspiro 3,3 heptane oxalic acid salt, 2-oxa-6-azaspiro 3.3 heptane xoxalate, 2-oxa-6-azaspiro 3.3 heptane oxalic acid, 2-oxa-6-azaspiro 3.3 heptane; oxalic acid, 2-oxa-6-azaspiro 3.3 heptane ethanedioate, 2-oxa-6-azaspiro 3.3 heptane oxalate 1:1, 2-oxa-6-azaspiro 3.3 heptane, ethanedioate 1:1, acmc-2099rjShow MoreShow Less
PubChem CID 53308464
SMILES OC(=O)C(O)=O.C1NCC11COC1

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
IUPAC Name 2-oxa-6-azaspiro[3.3]heptane;oxalic acid
Formula Weight 189.17
Percent Purity 97%
Chemical Name or Material 2-Oxa-6-azaspiro[3.3]heptane oxalate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberI13Z047 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberH63578.MD,H63578.06,H63578.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Oxa-6-azaspiro[3.3]heptane oxalate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.