missing translation for 'onlineSavingsMsg'
Learn More

2-Oxocyclohexanepropionitrile, 99%

Product Code. p-7033582
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11494630

Brand: Thermo Scientific Alfa Aesar H50722.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4594-78-9 | C9H13NO | 151.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4594-78-9
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD00013797
InChI Key SPTVCXKSSRBTMN-UHFFFAOYNA-N
Synonym 3-2-oxocyclohexyl propanenitrile, 2-2-cyanoethyl cyclohexanone, cyclohexanepropanenitrile, 2-oxo, 2-2-cyanethyl cyclohexanone, 2-oxo-1-cyclohexanepropionitrile, cyclohexanepropionitrile, 2-oxo, 2-oxocyclohexanepropionitrile, 2-.beta.-cyanoethyl cyclohexanone, 2-b-cyanoethyl cyclohexanone, 2-oxocyclohexanepropiononitrile
PubChem CID 96223
IUPAC Name 3-(2-oxocyclohexyl)propanenitrile
SMILES O=C1CCCCC1CCC#N

Specifications

Density 1.021
Boiling Point 138°C to 142°C (10 mmHg)
Quantity 1 g
UN Number UN3276
Formula Weight 151.21
Percent Purity 99%
Chemical Name or Material 2-Oxocyclohexanepropionitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.