missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Oxocyclopentylacetic acid, 97%, Thermo Scientific Chemicals
38.40 € - 131.00 €
Chemical Identifiers
CAS | 1460-38-4 |
---|---|
Molecular Formula | C7H10O3 |
Molecular Weight (g/mol) | 142.154 |
MDL Number | MFCD02093672 |
InChI Key | OLLLIBGOZUPLOK-UHFFFAOYSA-N |
Synonym | 2-2-oxocyclopentyl acetic acid, 2-oxocyclopentaneacetic acid, 2-oxo-cyclopentaneacetic acid, 2-oxocyclopentylacetic acid, 2-oxocyclopentyl acetic acid, 2-oxo-cyclopentaneaceticacid, acmc-1bx7z, 2-oxo-cyclopentyl acetic acid, cyclopentaneaceticacid, 2-oxo, 2-2-oxocyclopentyl aceticacid |
PubChem CID | 3569983 |
IUPAC Name | 2-(2-oxocyclopentyl)acetic acid |
SMILES | C1CC(C(=O)C1)CC(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11441600
|
Thermo Scientific Alfa Aesar
H26977.03 |
1 g |
38.40 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11451600
|
Thermo Scientific Alfa Aesar
H26977.06 |
5 g | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1460-38-4 | |
142.154 | |
OLLLIBGOZUPLOK-UHFFFAOYSA-N | |
3569983 | |
C1CC(C(=O)C1)CC(=O)O |
C7H10O3 | |
MFCD02093672 | |
2-2-oxocyclopentyl acetic acid, 2-oxocyclopentaneacetic acid, 2-oxo-cyclopentaneacetic acid, 2-oxocyclopentylacetic acid, 2-oxocyclopentyl acetic acid, 2-oxo-cyclopentaneaceticacid, acmc-1bx7z, 2-oxo-cyclopentyl acetic acid, cyclopentaneaceticacid, 2-oxo, 2-2-oxocyclopentyl aceticacid | |
2-(2-oxocyclopentyl)acetic acid |
Specifications
1460-38-4 | |
>110°C (230°F) | |
MFCD02093672 | |
OLLLIBGOZUPLOK-UHFFFAOYSA-N | |
2-(2-oxocyclopentyl)acetic acid | |
3569983 | |
97% |
47°C to 51°C | |
C7H10O3 | |
2-2-oxocyclopentyl acetic acid, 2-oxocyclopentaneacetic acid, 2-oxo-cyclopentaneacetic acid, 2-oxocyclopentylacetic acid, 2-oxocyclopentyl acetic acid, 2-oxo-cyclopentaneaceticacid, acmc-1bx7z, 2-oxo-cyclopentyl acetic acid, cyclopentaneaceticacid, 2-oxo, 2-2-oxocyclopentyl aceticacid | |
C1CC(C(=O)C1)CC(=O)O | |
142.154 | |
142.15 | |
2-Oxocyclopentylacetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only