missing translation for 'onlineSavingsMsg'
Learn More

2-Pentanone, 99+%, Thermo Scientific Chemicals

Artikelnummer. 11328006
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
500 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

2-Pentanone is used as a solvent for lacquers and surface coatings. It acts as a flavoring agent. It is also used as a solvent for cleaning and degreasing purpose. Further, it serves as an intermediate, paint additive and coating additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Pentanone is used as a solvent for lacquers and surface coatings. It acts as a flavoring agent. It is also used as a solvent for cleaning and degreasing purpose. Further, it serves as an intermediate, paint additive and coating additive.

Solubility
Miscible with water.

Notes
Incompatible with strong bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 107-87-9
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.134
MDL Number MFCD00009400
InChI Key XNLICIUVMPYHGG-UHFFFAOYSA-N
Synonym 2-pentanone, methyl propyl ketone, ethyl acetone, pentanone, ethylacetone, propyl methyl ketone, metylopropyloketon, methyl n-propyl ketone, methylpropyl ketone, methyl-propyl-cetone
PubChem CID 7895
ChEBI CHEBI:16472
IUPAC Name pentan-2-one
SMILES CCCC(=O)C

Spezifikation

Melting Point -78°C
Density 0.813
Boiling Point 100°C to 101°C
Flash Point 7°C (44°F)
Refractive Index 1.3905
Quantity 25 g
UN Number UN1249
Beilstein 506058
Merck Index 14,6114
Solubility Information Miscible with water.
Formula Weight 86.13
Percent Purity ≥99%
Chemical Name or Material 2-Pentanone
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons
Identificativo prodotto
  • 2-Pentanone
Avvertenza
  • Danger
Categoria di pericolo
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Indicazioni di pericolo
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
Consigli di prudenza
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Informazioni supplementari
  • MIXTURE LIST-Contains : Methyl propyl ketone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'