missing translation for 'onlineSavingsMsg'
Learn More

2-Pentyn-1-ol, 98%, Thermo Scientific Chemicals

Product Code. 11449893
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6261-22-9
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00040915
InChI Key WLPYSOCRPHTIDZ-UHFFFAOYSA-N
Synonym 2-pentyn-1-ol, 2-pentyne-1-ol, 2-pentynol, zlchem 1280, pubchem13063, acmc-209n5w, ksc357i0r, 2-pentyn-1-01
PubChem CID 80421
IUPAC Name pent-2-yn-1-ol
SMILES CCC#CCO

Specifications

Density 0.91
Boiling Point 154°C
Flash Point 58°C (136°F)
Odor Recognizable
Linear Formula CH3CH2C≡CCH2OH
Refractive Index 1.453
Quantity 5 g
UN Number UN1987
Beilstein 1734007
Formula Weight 84.12
Percent Purity 98%
Chemical Name or Material 2-Pentyn-1-ol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'