missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenoxyethyl bromide, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
It is used in the synthesis and antihypertensive activity of 4'-substituted spiro[4H-3,1-benzoxazine-4,4'-piperidin]-2(1H)-ones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the synthesis and antihypertensive activity of 4′-substituted spiro[4H-3,1-benzoxazine-4,4′-piperidin]-2(1H)-ones.
Solubility
Insoluble in water. Solubility in methanol is almost transparent. Soluble in chloroform and ethyl acetate.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is used in the synthesis and antihypertensive activity of 4′-substituted spiro[4H-3,1-benzoxazine-4,4′-piperidin]-2(1H)-ones.
Solubility
Insoluble in water. Solubility in methanol is almost transparent. Soluble in chloroform and ethyl acetate.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
Melting Point | 31°C to 35°C |
Density | 1.35 |
Boiling Point | 135°C to 137°C (20 mmHg) |
Flash Point | 65°C (149°F) |
Quantity | 25 g |
Beilstein | 508290 |
Solubility Information | Insoluble in water. Solubility in methanol is almost transparent. Soluble in chloroform and ethyl acetate. |
Formula Weight | 201.07 |
Percent Purity | 98% |
Chemical Name or Material | 2-Phenoxyethyl bromide |
Safety and Handling
Product Identifier
- 2-Phenoxyethyl bromide
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : (2-bromoethoxy)benzene
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction