missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxyethylamine, 98%, Thermo Scientific Chemicals

37.59 € - 463.31 €

Chemical Identifiers

CAS 1758-46-9
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00052975
InChI Key IMLAIXAZMVDRGA-UHFFFAOYSA-N
Synonym 2-phenoxyethylamine, phenoxyethylamine, ethanamine, 2-phenoxy, 2-phenoxylethylamine, 2-phenoxy-ethylamine, 2-aminoethoxy benzene, ethylamine, 2-phenoxy, alpha-phenoxy-beta-aminoethane, unii-8dgq1b38r5, 2-phenoxyethylamino
PubChem CID 15651
IUPAC Name 2-phenoxyethanamine
SMILES C1=CC=C(C=C1)OCCN
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11474903
View Documents
Thermo Scientific Alfa Aesar
A12225.03
1 g
37.59 €
1g
Please to purchase this item. Need a web account? Register with us today!
11484903
View Documents
Thermo Scientific Alfa Aesar
A12225.06
5 g
115.58 €
5g
Please to purchase this item. Need a web account? Register with us today!
11494903
View Documents
Thermo Scientific Alfa Aesar
A12225.14
25 g
463.31 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Organic intermediate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1758-46-9
137.182
IMLAIXAZMVDRGA-UHFFFAOYSA-N
15651
C1=CC=C(C=C1)OCCN
C8H11NO
MFCD00052975
2-phenoxyethylamine, phenoxyethylamine, ethanamine, 2-phenoxy, 2-phenoxylethylamine, 2-phenoxy-ethylamine, 2-aminoethoxy benzene, ethylamine, 2-phenoxy, alpha-phenoxy-beta-aminoethane, unii-8dgq1b38r5, 2-phenoxyethylamino
2-phenoxyethanamine

Specifications

1758-46-9
107°C (224°F)
C8H11NO
MFCD00052975
774671
IMLAIXAZMVDRGA-UHFFFAOYSA-N
2-phenoxyethanamine
15651
98%
88°C to 90°C (1 mmHg)
Amine-like
C6H5O(CH2)2NH2
UN2735
2-phenoxyethylamine, phenoxyethylamine, ethanamine, 2-phenoxy, 2-phenoxylethylamine, 2-phenoxy-ethylamine, 2-aminoethoxy benzene, ethylamine, 2-phenoxy, alpha-phenoxy-beta-aminoethane, unii-8dgq1b38r5, 2-phenoxyethylamino
C1=CC=C(C=C1)OCCN
137.182
137.18
2-Phenoxyethylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P308+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or concerned: Get medical advice/attention.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

missing translation for 'rtecsNumber' : KR9500000

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.