missing translation for 'onlineSavingsMsg'
Learn More

(2-Phenyl-1,3-thiazol-4-yl)methylamine, 97%, Thermo Scientific™

Product Code. 10079231
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 165736-03-8
Molecular Formula C10H10N2S
Molecular Weight (g/mol) 190.264
MDL Number MFCD02682055
InChI Key PMFCMLGAPHZRTE-UHFFFAOYSA-N
Synonym 2-phenyl-1,3-thiazol-4-yl methylamine, 2-phenylthiazol-4-yl methanamine, 2-phenyl-1,3-thiazol-4-yl methanamine, c-2-phenyl-thiazol-4-yl-methylamine, 1-2-phenyl-1,3-thiazol-4-yl methanamine, 4-thiazolemethanamine, 2-phenyl, 4-aminomethyl-2-phenyl-1,3-thiazole, enamine_005991, 2-phenyl-1,3-thiazol-4-yl methanamine dihydrochloride, chembrdg-bb 4101065
PubChem CID 736526
IUPAC Name (2-phenyl-1,3-thiazol-4-yl)methanamine
SMILES C1=CC=C(C=C1)C2=NC(=CS2)CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 190.27
Percent Purity 97%
Chemical Name or Material (2-Phenyl-1, 3-thiazol-4-yl)methylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'