missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenyl-2-butenal, (E)+(Z), 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
2-Phenyl-2-butenal is used as a food additive. It is also used as intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Phenyl-2-butenal is used as a food additive. It is also used as intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in oil and ethanol. Insoluble in water.
Notes
Air Sensitive. Incompatible with oxidizing agents and reducing agents.
2-Phenyl-2-butenal is used as a food additive. It is also used as intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in oil and ethanol. Insoluble in water.
Notes
Air Sensitive. Incompatible with oxidizing agents and reducing agents.
Specifications
Specifications
Boiling Point | 115°C (15 mmHg) |
Refractive Index | 1.561 |
Quantity | 25 g |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in oil and ethanol. Insoluble in water. |
Formula Weight | 146.19 |
Percent Purity | 97% |
Chemical Name or Material | 2-Phenyl-2-butenal, (E)+(Z) |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'