missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-2-pentenal, (E)+(Z), 90+%, Thermo Scientific Chemicals

Product Code. 11420214
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3491-63-2
Molecular Formula C11H12O
Molecular Weight (g/mol) 160.216
MDL Number MFCD00051835
InChI Key YPAJRUMMODCONM-IZZDOVSWSA-N
Synonym 2-phenyl-2-pentenal, 2z-2-phenylpent-2-enal
PubChem CID 15310469
IUPAC Name (Z)-2-phenylpent-2-enal
SMILES CCC=C(C=O)C1=CC=CC=C1

Specifications

Density 1.005
Boiling Point 124°C to 126°C (14 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.551
Quantity 5 g
Beilstein 1859032
Sensitivity Air sensitive
Formula Weight 160.22
Percent Purity ≥90%
Chemical Name or Material 2-Phenyl-2-pentenal, (E)+(Z)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.