missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenylacetamide, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
Description
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a pharmaceutical intermediate.
Solubility
Soluble in methanol.
Notes
Store at 4°C.
It is used as a pharmaceutical intermediate.
Solubility
Soluble in methanol.
Notes
Store at 4°C.

Specifications
Specifications
CAS | 103-81-1 |
Molecular Formula | C8H9NO |
MDL Number | MFCD00059193 |
Quantity | 250 mg |
Synonym | benzeneacetamide, phenylacetamide, alpha-toluamide, alpha-phenylacetamide, phenylacetic acid amide, acetamide, 2-phenyl, phenyl-beta-acetylamine, alpha-toluimidic acid, 2-phenyl-acetamide, phenyl acetamide |
Solubility Information | Soluble in methanol. |
InChI Key | LSBDFXRDZJMBSC-UHFFFAOYSA-N |
SMILES | NC(=O)CC1=CC=CC=C1 |
IUPAC Name | 2-phenylacetamide |
Molecular Weight (g/mol) | 135.17 |
Show More |
Safety and Handling

Product Identifier
- 2-Phenylacetamide
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- P264-Wash face, hands, and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P330-Rinse mouth.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P403-Store in a well-ventilated place.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Benzeneacetamide
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction