missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylmalonamide, 97%, Thermo Scientific Chemicals

Product Code. 11451537
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10255-95-5
Molecular Formula C9H10N2O2
Molecular Weight (g/mol) 178.19
MDL Number MFCD00051776
InChI Key CPSUAFUQJBJMPO-UHFFFAOYSA-N
Synonym 2-phenylmalonamide, propanediamide, 2-phenyl, phenylmethane-1,1-dicarboxamide, phenylmalonamide, 2-phenyl-malonamide, 2-phenylmalondiamide, acmc-20anni, maybridge1_000163, ksc492s5h
PubChem CID 2747536
IUPAC Name 2-phenylpropanediamide
SMILES NC(=O)C(C(N)=O)C1=CC=CC=C1

Specifications

Melting Point ∼233°C (decomposition)
Quantity 1 g
Beilstein 2446518
Formula Weight 178.19
Percent Purity 97%
Chemical Name or Material 2-Phenylmalonamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'