missing translation for 'onlineSavingsMsg'
Learn More
2-Propanol, BAKER ANALYZED, J.T. Baker™ Product Code.: 10454421
compliance-icons compliance-icons compliance-icons

2-Propanol, BAKER ANALYZED, J.T. Baker™

Product Code. 10454421
5 L, EcoTainer Metal Safety Can
Click to view available options
Quantity:
5 L
Packaging:
EcoTainer Metal Safety Can
This item is not returnable. View return policy

Product Code. 10454421

Brand: JT Baker 8067.5000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High quality grade chemical that exceeds ACS specifications for laboratory and specialized industrial use.

  • Exceeds the requirements set forth by the American Chemical Society (ACS)
  • Label provides an actual Lot Analysis as a guarantee of purity and consistency
  • Analytical methods used for these determinations are based on standard test procedures described in technical publications
  • Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability

Specifications

Melting Point -89.5°C
CAS 67-63-0
Density 0.79g/cm3
Boiling Point 82°C
Quantity 5 L
Flash Point 12°C
Ignition Point 425°C
Odor Alcohol-like
Packaging EcoTainer Metal Safety Can
Physical Form Liquid
Residue after Evaporation ≤0.001%
Molecular Formula C3H8O
Linear Formula CH3CHOHCH3
Viscosity 2.43 mPas
Water ≤0.1%
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILES CC(C)O
IUPAC Name propan-2-ol
Molecular Weight (g/mol) 60.096
PubChem CID 3776
ChEBI CHEBI:17824
Formula Weight 60.1g/mol
Percent Purity ≥99.5%
Grade Exceeds ACS specifications
Chemical Name or Material 2-Propanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Propanol, BAKER ANALYZED, J.T. Baker™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.