missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, Laboratory Reagent, ≥99.5%, Honeywell™

Product Code. 15660990
Click to view available options
Quantity:
1 L
2.5 L
10 L
20 L
25 L
4 x 2.5 L
Packaging:
Glass Bottle
Stainless Steel Can

Laboratory Reagent,≥99.5%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point -90°C
Color Colorless
Density 0.785g/mL at 25°C
Boiling Point 77°C to 83°C (1013 hPa)
Quantity 2.5 L
Explosion Limit 0.02
Flash Point 12°C (53.6°F)
Ignition Point 750°F
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.377(lit.)
Linear Formula (CH3)2CHOH
Vapor Pressure 33mmHg (20°C), 44mmHg (25°C)
UN Number UN1219
Beilstein 635639
Solubility Information Soluble in water
Formula Weight 60.1g/mol
Percent Purity ≥99.5%
Grade Laboratory Reagent
Chemical Name or Material 2-Propanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Propanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'