missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propanol, CHROMASOLV™ LC-MS, Honeywell Riedel-de Haën™
CHROMASOLV™ LC-MS
82.41 € - 567.59 €
Chemical Identifiers
CAS | 67-63-0 |
---|---|
Molecular Formula | C3H8O |
Molecular Weight (g/mol) | 60.096 |
MDL Number | MFCD00011674 |
InChI Key | KFZMGEQAYNKOFK-UHFFFAOYSA-N |
Synonym | isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol |
PubChem CID | 3776 |
ChEBI | CHEBI:17824 |
IUPAC Name | propan-2-ol |
SMILES | CC(C)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15664730
|
Honeywell-Chromasolv
34965-1L |
1 L |
82.41 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15674730
|
Honeywell-Chromasolv
34965-2.5L |
2.5 L |
159.82 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15694730
|
Honeywell-Chromasolv
34965-6X1L |
6 x 1 L |
426.89 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15684730
|
Honeywell-Chromasolv
34965-4X2.5L |
4 x 2.5 L |
567.59 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- For LC-MS and LC-UV trace metals analysis
- HPLC / UV-VIS quality control
- Low gradient baseline drift
- Low levels of trace metals
- No non-volatile impurities
Chemical Identifiers
67-63-0 | |
60.096 | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
3776 | |
propan-2-ol |
C3H8O | |
MFCD00011674 | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
CHEBI:17824 | |
CC(C)O |
Specifications
67-63-0 | |
0.785g/mL at 25°C | |
12°C (53.6°F) | |
C3H8O | |
MFCD00011674 | |
UN1219 | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
CC(C)O | |
60.096 | |
CHEBI:17824 | |
≥99.9% | |
2-Propanol |
Colorless | |
0.02 | |
750°F | |
(CH3)2CHOH | |
44 mmHg (25°C), 33 mmHg (20°C) | |
635639 | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
propan-2-ol | |
3776 | |
60.1g/mol | |
LC-MS CHROMASOLV™ |
Safety and Handling
P210-P261-P305 + P351 + P338
missing translation for 'rtecsNumber' : NT8050000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 2-Propanol
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : propan-2-ol