missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, Puriss. p.a., ACS Reagent, ≥99.8% (GC), Honeywell Riedel-de Haën™

Product Code. 15615890
Click to view available options
Quantity:
4 x 25 mL
4 x 2.5 L
6 x 1 L
155 kg
Packaging:
Glass Bottle
Metal Drum
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss. p.a., ACS Reagent,≥99.8% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Density 0.785g/mL at 20°C, 0.785g/mL at 25°C
Boiling Point 77°C to 83°C
Quantity 155 kg
Explosion Limit 0.02
Packaging Metal Drum
Refractive Index 1.378
Linear Formula (CH3)2CHOH
UN Number UN1219
Viscosity 4.3 mPas at 20°C
Beilstein 635639
Molecular Weight (g/mol) 60.096
Formula Weight 60.1g/mol
Percent Purity ≥99.8% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material 2-Propanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Propanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'