missing translation for 'onlineSavingsMsg'
Learn More
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., Honeywell Product Code.: 15653990

2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., Honeywell

Product Code. 15653990
1 L, Plastic Bottle
Click to view available options
Quantity:
1 L
2.5 L
5 L
10 L
25 L
155 kg
Packaging:
Glass Bottle
Metal Drum
Plastic Bottle
Stainless Steel Can
This item is not returnable. View return policy

Product Code. 15653990

Brand: Honeywell Riedel-de Haën 335391L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petroholShow MoreShow Less
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Density 0.785g/mL at 25°C
Quantity 1 L
Explosion Limit 0.02
Flash Point 12°C (53.6°F)
Ignition Point 750°F
Packaging Plastic Bottle
Linear Formula (CH3)2CHOH
Vapor Pressure 33 mmHg (20°C), 44 mmHg (25°C)
UN Number UN1219
Beilstein 635639
Formula Weight 60.1g/mol
Percent Purity ≥99.8% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material 2-Propanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Propanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., Honeywell >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.