missing translation for 'onlineSavingsMsg'
Learn More

2-Propionylthiophene, 98%, Thermo Scientific Chemicals

Product Code. 11337665
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13679-75-9
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.2
MDL Number MFCD00005446
InChI Key MFPZQZZWAMAHOY-UHFFFAOYSA-N
Synonym 1-2-thienyl-1-propanone, 2-propionylthiophene, 1-thiophen-2-yl propan-1-one, 1-2-thienyl propan-1-one, ethyl 2-thienyl ketone, 2-propanoylthiophene, 1-propanone, 1-2-thienyl, unii-ny658tvp5m, ethyl thienyl ketone, ethyl-2-thienyl ketone
PubChem CID 26179
IUPAC Name 1-thiophen-2-ylpropan-1-one
SMILES CCC(=O)C1=CC=CS1

Specifications

Density 1.124
Boiling Point 228°C
Flash Point 100°C (212°F)
Refractive Index 1.553
Quantity 25 g
Beilstein 108934
Formula Weight 140.2
Percent Purity 98%
Chemical Name or Material 2-Propionylthiophene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'