missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrazinecarbonyl chloride, Technical Grade, Thermo Scientific™

Product Code. 10049081
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 19847-10-0
Molecular Formula C5H3ClN2O
Molecular Weight (g/mol) 142.542
InChI Key TXJKATOSKLUITR-UHFFFAOYSA-N
Synonym 2-pyrazinecarbonyl chloride, pyrazinecarbonyl chloride, pyrazine-2-carboxylic acid chloride, 2-pyrazinecarbonylchloride, pyrazine 2-carbonyl chloride, 2-pyrazine carbonyl chloride, pyrazinoyl chloride, acmc-20ann0, 2-pyrazine carboxylic acid chloride, pyrazine-2-carbonyl chloride, technical 1g
PubChem CID 88279
IUPAC Name pyrazine-2-carbonyl chloride
SMILES C1=CN=C(C=N1)C(=O)Cl

Specifications

Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 142.54
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-Pyrazinecarbonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'