missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Pyridin-3-ylbenzaldehyde, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Quantity | 10 g |
Synonym | 2-pyridin-3-yl benzaldehyde, 2-pyridin-3-yl-benzaldehyde, 2-3-pyridinyl benzaldehyde, 2-3-pyridyl benzaldehyde, benzaldehyde,2-3-pyridinyl, 2-3-pyrldinyl benzaldehyde, benzaldehyde, 2-3-pyridinyl, pubchem22510, dtuanrrvvjrtjs-uhfffaoysa |
Formula Weight | 183.21 |
Percent Purity | 97% |
Chemical Name or Material | 2-Pyridin-3-ylbenzaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'