Learn More
2-Pyridin-3-ylpiperidine, 97%, Thermo Scientific Chemicals
236.18 € - 711.54 €
Chemical Identifiers
CAS | 13078-04-1 |
---|---|
Molecular Formula | C10H14N2 |
Molecular Weight (g/mol) | 162.24 |
MDL Number | MFCD00006370 |
InChI Key | MTXSIJUGVMTTMU-UHFFFAOYNA-N |
Synonym | 3-piperidin-2-yl pyridine, anabasine, dl-anabasine, neonicotine, +--anabasine, 2-pyridin-3-ylpiperidine, +/--anabasine, 3-2-piperidinyl pyridine, 2-3-pyridyl piperidine, pyridine, 3-2-piperidinyl |
PubChem CID | 2181 |
ChEBI | CHEBI:28986 |
IUPAC Name | 3-piperidin-2-ylpyridine |
SMILES | C1CCC(NC1)C1=CC=CN=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15552353
|
Thermo Scientific Acros
458980010 |
1 g |
236.18 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15562353
|
Thermo Scientific Acros
458980050 |
5 g |
711.54 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13078-04-1 | |
162.24 | |
MTXSIJUGVMTTMU-UHFFFAOYNA-N | |
2181 | |
3-piperidin-2-ylpyridine |
C10H14N2 | |
MFCD00006370 | |
3-piperidin-2-yl pyridine, anabasine, dl-anabasine, neonicotine, +--anabasine, 2-pyridin-3-ylpiperidine, +/--anabasine, 3-2-piperidinyl pyridine, 2-3-pyridyl piperidine, pyridine, 3-2-piperidinyl | |
CHEBI:28986 | |
C1CCC(NC1)C1=CC=CN=C1 |
Specifications
13078-04-1 | |
1.0460g/mL | |
93°C | |
96% min. (GC) | |
C10H14N2 | |
1.046 | |
MTXSIJUGVMTTMU-UHFFFAOYNA-N | |
3-piperidin-2-ylpyridine | |
2181 | |
162.23 | |
2-Pyridin-3-ylpiperidine |
9.0°C | |
270.0°C to 272.0°C | |
Authentic | |
Glass Bottle | |
MFCD00006370 | |
3-piperidin-2-yl pyridine, anabasine, dl-anabasine, neonicotine, +--anabasine, 2-pyridin-3-ylpiperidine, +/--anabasine, 3-2-piperidinyl pyridine, 2-3-pyridyl piperidine, pyridine, 3-2-piperidinyl | |
C1CCC(NC1)C1=CC=CN=C1 | |
162.24 | |
CHEBI:28986 | |
97% |
Safety and Handling
GHS H Statement
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
Danger
- 2-Pyridin-3-ylpiperidine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H300-Fatal if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.