Learn More
2-Pyridinemethanol, 98+%, Thermo Scientific Chemicals
36.28 € - 433.16 €
Chemical Identifiers
CAS | 586-98-1 |
---|---|
Molecular Formula | C6H7NO |
Molecular Weight (g/mol) | 109.128 |
MDL Number | MFCD00006348 |
InChI Key | SHNUBALDGXWUJI-UHFFFAOYSA-N |
Synonym | 2-pyridinemethanol, 2-hydroxymethyl pyridine, piconol, 2-pyridinylmethanol, 2-pyridylmethanol, 2-pyridylcarbinol, pyridine-2-methanol, pyridine-2-carbinol, alpha-picolyl alcohol, pyridin-2-yl methanol |
PubChem CID | 11474 |
IUPAC Name | pyridin-2-ylmethanol |
SMILES | C1=CC=NC(=C1)CO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11497227
|
Thermo Scientific Alfa Aesar
A15005.14 |
25 g |
36.28 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11407237
|
Thermo Scientific Alfa Aesar
A15005.22 |
100 g |
101.51 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11417237
|
Thermo Scientific Alfa Aesar
A15005.36 |
500 g |
433.16 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Pyridinemethanol is used as a pyridylalcohol with hypoglycemic activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Pyridinemethanol is used as a pyridylalcohol with hypoglycemic activity.
Solubility
Fully miscible in water.
Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
586-98-1 | |
109.128 | |
SHNUBALDGXWUJI-UHFFFAOYSA-N | |
11474 | |
C1=CC=NC(=C1)CO |
C6H7NO | |
MFCD00006348 | |
2-pyridinemethanol, 2-hydroxymethyl pyridine, piconol, 2-pyridinylmethanol, 2-pyridylmethanol, 2-pyridylcarbinol, pyridine-2-methanol, pyridine-2-carbinol, alpha-picolyl alcohol, pyridin-2-yl methanol | |
pyridin-2-ylmethanol |
Specifications
586-98-1 | |
1.131 | |
120°C (248°F) | |
1.544 | |
107849 | |
2-pyridinemethanol, 2-hydroxymethyl pyridine, piconol, 2-pyridinylmethanol, 2-pyridylmethanol, 2-pyridylcarbinol, pyridine-2-methanol, pyridine-2-carbinol, alpha-picolyl alcohol, pyridin-2-yl methanol | |
SHNUBALDGXWUJI-UHFFFAOYSA-N | |
pyridin-2-ylmethanol | |
11474 | |
≥98% |
-6°C to -5°C | |
112°C to 113°C (16 mmHg) | |
C6H7NO | |
MFCD00006348 | |
Hygroscopic | |
Fully miscible in water. | |
C1=CC=NC(=C1)CO | |
109.128 | |
109.13 | |
2-Pyridinemethanol |
Safety and Handling
GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
H302-H315-H319
missing translation for 'einecsNumber' : 209-592-7
missing translation for 'rtecsNumber' : UT4689000
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.