missing translation for 'onlineSavingsMsg'
Learn More

2-Pyridylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Product Code. 15465375
Click to view available options
Quantity:
50 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the preparation of P,N ligands for asymmetric Ir-catalyzed reactions.

Solubility
Reacts with water.

Notes
Handle under dry protective gas. Air sensitive. Store away from strong bases a
TRUSTED_SUSTAINABILITY

Specifications

CAS 218777-23-2
Density 0.974
Molecular Formula C5H5BrNZn-
MDL Number MFCD00671985
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive; May form a precipitate
Synonym 2-pyridylzinc bromide
Solubility Information Reacts with water.
InChI Key WUTSMSLAOBVVCP-UHFFFAOYSA-M
SMILES C1=CC=NC=C1.[Zn].[Br-]
IUPAC Name pyridine;zinc;bromide
Molecular Weight (g/mol) 224.386
PubChem CID 76056926
Formula Weight 223.38
Chemical Name or Material 2-Pyridylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'