missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrrolidin-1-yl-1,3-thiazole-5-carbaldehyde, 97%, Thermo Scientific™

41.90 € - 78.90 €

Chemical Identifiers

CAS 900015-48-7
Molecular Formula C8H10N2OS
Molecular Weight (g/mol) 182.241
MDL Number MFCD07021330
InChI Key CFFWKLLEEVWNLE-UHFFFAOYSA-N
Synonym 2-1-pyrrolidinyl-1,3-thiazole-5-carbaldehyde, 2-pyrrolidin-1-yl-1,3-thiazole-5-carbaldehyde, 2-pyrrolidinyl-1,3-thiazole-5-carbaldehyde, 2-pyrrolidin-1-yl thiazole-5-carbaldehyde
PubChem CID 2767663
IUPAC Name 2-pyrrolidin-1-yl-1,3-thiazole-5-carbaldehyde
SMILES C1CCN(C1)C2=NC=C(S2)C=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10153563
View Documents
Thermo Scientific Maybridge
CC64204CB
250 mg Amber Glass Bottle
41.90 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10103414
View Documents
Thermo Scientific Maybridge
CC64204DA
1 g Amber Glass Bottle
78.90 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

900015-48-7
182.241
CFFWKLLEEVWNLE-UHFFFAOYSA-N
2767663
C1CCN(C1)C2=NC=C(S2)C=O
C8H10N2OS
MFCD07021330
2-1-pyrrolidinyl-1,3-thiazole-5-carbaldehyde, 2-pyrrolidin-1-yl-1,3-thiazole-5-carbaldehyde, 2-pyrrolidinyl-1,3-thiazole-5-carbaldehyde, 2-pyrrolidin-1-yl thiazole-5-carbaldehyde
2-pyrrolidin-1-yl-1,3-thiazole-5-carbaldehyde

Specifications

900015-48-7
100.0
C8H10N2OS
2-1-pyrrolidinyl-1,3-thiazole-5-carbaldehyde, 2-pyrrolidin-1-yl-1,3-thiazole-5-carbaldehyde, 2-pyrrolidinyl-1,3-thiazole-5-carbaldehyde, 2-pyrrolidin-1-yl thiazole-5-carbaldehyde
C1CCN(C1)C2=NC=C(S2)C=O
182.241
182.25
2-Pyrrolidin-1-yl-1, 3-thiazole-5-carbaldehyde
96.5
97%
MFCD07021330
CFFWKLLEEVWNLE-UHFFFAOYSA-N
2-pyrrolidin-1-yl-1,3-thiazole-5-carbaldehyde
2767663
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Product Certifications