Learn More
2-Pyrrolidin-1-ylisonicotinaldehyde, 97%, Thermo Scientific™
98.20 € - 280.00 €
Chemical Identifiers
CAS | 898289-23-1 |
---|---|
Molecular Formula | C10H12N2O |
Molecular Weight (g/mol) | 176.219 |
MDL Number | MFCD08690286 |
InChI Key | QPKFMHRNVDJMLZ-UHFFFAOYSA-N |
Synonym | 2-pyrrolidin-1-ylisonicotinaldehyde, 2-pyrrolidin-1-yl isonicotinaldehyde, 2-pyrrolidin-1-yl pyridine-4-carbaldehyde, 2-pyrrolidin-1-yl pyridine-4-carboxaldehyde, 4-pyridinecarboxaldehyde,2-1-pyrrolidinyl, 2-pyrrolidinylpyridine-4-carbaldehyde, 2-pyrrolidin-1-yl-isonicotinaldehyde, 4-pyridinecarboxaldehyde, 2-1-pyrrolidinyl |
PubChem CID | 18525845 |
IUPAC Name | 2-pyrrolidin-1-ylpyridine-4-carbaldehyde |
SMILES | C1CCN(C1)C2=NC=CC(=C2)C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10526324
|
Thermo Scientific Maybridge
CC50404CB |
250 mg | Amber Glass Bottle |
98.20 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10631994
|
Thermo Scientific Maybridge
CC50404DA |
1 g | Amber Glass Bottle |
280.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
898289-23-1 | |
176.219 | |
QPKFMHRNVDJMLZ-UHFFFAOYSA-N | |
18525845 | |
C1CCN(C1)C2=NC=CC(=C2)C=O |
C10H12N2O | |
MFCD08690286 | |
2-pyrrolidin-1-ylisonicotinaldehyde, 2-pyrrolidin-1-yl isonicotinaldehyde, 2-pyrrolidin-1-yl pyridine-4-carbaldehyde, 2-pyrrolidin-1-yl pyridine-4-carboxaldehyde, 4-pyridinecarboxaldehyde,2-1-pyrrolidinyl, 2-pyrrolidinylpyridine-4-carbaldehyde, 2-pyrrolidin-1-yl-isonicotinaldehyde, 4-pyridinecarboxaldehyde, 2-1-pyrrolidinyl | |
2-pyrrolidin-1-ylpyridine-4-carbaldehyde |
Specifications
898289-23-1 | |
100.0 | |
C10H12N2O | |
2-pyrrolidin-1-ylisonicotinaldehyde, 2-pyrrolidin-1-yl isonicotinaldehyde, 2-pyrrolidin-1-yl pyridine-4-carbaldehyde, 2-pyrrolidin-1-yl pyridine-4-carboxaldehyde, 4-pyridinecarboxaldehyde,2-1-pyrrolidinyl, 2-pyrrolidinylpyridine-4-carbaldehyde, 2-pyrrolidin-1-yl-isonicotinaldehyde, 4-pyridinecarboxaldehyde, 2-1-pyrrolidinyl | |
C1CCN(C1)C2=NC=CC(=C2)C=O | |
176.219 | |
176.22 | |
2-Pyrrolidin-1-ylisonicotinaldehyde |
96.5 | |
97% | |
MFCD08690286 | |
QPKFMHRNVDJMLZ-UHFFFAOYSA-N | |
2-pyrrolidin-1-ylpyridine-4-carbaldehyde | |
18525845 | |
97% |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
Toxic if inhaled.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Toxic in contact with skin.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It
GHS Signal Word: Danger
Your input is important to us. Please complete this form to provide feedback related to the content on this product.