missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrrolidin-2-ylpyridine, 95%, Thermo Scientific Chemicals

Product Code. 13545000
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77790-61-5
Molecular Formula C9H12N2
Molecular Weight (g/mol) 148.21
InChI Key NDCZQFDBSPOUDF-UHFFFAOYSA-N
Synonym 2-pyrrolidin-2-yl pyridine, 2-2-pyrrolidinyl pyridine, 2-pyrrolidin-2-yl-pyridine, +--alpha-nornicotine, pyridine, 2-2-pyrrolidinyl, +--2-2-pyrrolidinyl pyridine, pyridine, 2-2-pyrrolidinyl-, +-, r-2-2-pyrrolidinyl-pyridine, alpha-nornicotine, 2,3'-pyridylpyrrolidine
PubChem CID 2771659
IUPAC Name 2-pyrrolidin-2-ylpyridine
SMILES C1CC(NC1)C2=CC=CC=N2

Specifications

Boiling Point 82.0°C (0.8 mmHg)
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 148.21
Percent Purity 95%
Chemical Name or Material 2-Pyrrolidin-2-ylpyridine
compliance-icons
Product Identifier
  • (±)-2-(2-Pyrrolidinyl)pyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.